Assay Details
Gene Symbol: MEAK7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003126827.4 | XP_003126875.1 | 510 | 98 | ||
XM_005655749.3 | XP_005655806.1 | 510 | 98 | |||
XM_013988477.2 | XP_013843931.1 | 602 | 98 | |||
XM_013988478.2 | XP_013843932.1 | 602 | 98 | |||
XM_013988480.2 | XP_013843934.1 | 1349 | 98 | |||
XM_013988482.2 | XP_013843936.1 | 602 | 98 | |||
XM_021093727.1 | XP_020949386.1 | 976 | 98 |
