Assay Details
Gene Symbol: MEAK7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003126827.4 | XP_003126875.1 | 1202 | 68 | ||
XM_013988478.2 | XP_013843932.1 | 1294 | 68 | |||
XM_013988480.2 | XP_013843934.1 | 2041 | 68 | |||
XM_013988482.2 | XP_013843936.1 | 1294 | 68 | |||
XM_021093727.1 | XP_020949386.1 | 1668 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() |
