Assay Details
Gene Symbol: DYNC2I1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021079024.1 | XP_020934683.1 | 3609 | 76 | ||
XM_021079025.1 | XP_020934684.1 | 3596 | 76 | |||
XM_021079026.1 | XP_020934685.1 | 3608 | 76 | |||
XM_021079027.1 | XP_020934686.1 | 3596 | 76 |
