Assay Details
| Gene Symbol: MAFK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003124249.6 | XP_003124297.1 | 211 | 119 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_003124249.6 transcripts.
Set Membership: |
|

MAFK
Genomic Map