Assay Details
Gene Symbol: CBFA2T2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005672844.3 | XP_005672901.1 | 1428 | 72 | ||
XM_005672845.3 | XP_005672902.1 | 1100 | 72 | |||
XM_013985268.2 | XP_013840722.1 | 1467 | 72 | |||
XM_021078106.1 | XP_020933765.1 | 1210 | 72 | |||
XM_021078107.1 | XP_020933766.1 | 1198 | 72 | |||
XM_021078108.1 | XP_020933767.1 | 1260 | 72 | |||
XM_021078109.1 | XP_020933768.1 | 3751 | 72 | |||
XM_021078110.1 | XP_020933769.1 | 1092 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
