Assay Details
| Gene Symbol: YAF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021092540.1 | XP_020948199.1 | 639 | 74 | ||
| XM_021092541.1 | XP_020948200.1 | 700 | 74 | |||
| XM_021092542.1 | XP_020948201.1 | 778 | 74 | |||
| XM_021092543.1 | XP_020948202.1 | 705 | 74 | |||
| XM_021092544.1 | XP_020948203.1 | 567 | 74 | |||
| XM_021092545.1 | XP_020948204.1 | 3191 | 74 | |||
More Information
Set Membership: |
|

YAF2
Genomic Map