Assay Details
| Gene Symbol: CENPC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003482404.4 | XP_003482452.2 | 634 | 76 | ||
| XM_013978751.2 | XP_013834205.2 | 571 | 76 | |||
| XM_021101386.1 | XP_020957045.1 | 678 | 76 | |||
More Information
Set Membership: |
|

CENPC
Genomic Map