Assay Details
Gene Symbol: CYTIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003483653.4 | XP_003483701.1 | 390 | 90 | ||
XM_021074748.1 | XP_020930407.1 | 6437 | 90 | |||
XM_021074749.1 | XP_020930408.1 | 368 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() |
