Assay Details
Gene Symbol: ADGRF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013996166.2 | XP_013851620.1 | 355 | 83 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: ADGRF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013996166.2 | XP_013851620.1 | 355 | 83 |