Assay Details
Gene Symbol: LOC100513450 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003135085.4 | XP_003135133.1 | 412 | 103 | ||
XM_003135086.3 | XP_003135134.1 | 429 | 103 | |||
XM_005673619.2 | XP_005673676.1 | 405 | 103 | |||
XM_005673620.2 | XP_005673677.1 | 409 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() |
