Assay Details
| Gene Symbol: ASB14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021068997.1 | XP_020924656.1 | 2266 | 96 | ||
| XM_021068998.1 | XP_020924657.1 | 2267 | 96 | |||
| XM_021068999.1 | XP_020924658.1 | 2282 | 96 | |||
| XM_021069001.1 | XP_020924660.1 | 1930 | 96 | |||
| XM_021069002.1 | XP_020924661.1 | 1945 | 96 | |||
| XM_021069003.1 | XP_020924662.1 | 1684 | 96 | |||
More Information
Set Membership: |
|

ASB14
Genomic Map