Assay Details
| Gene Symbol: NPFFR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021101419.1 | XP_020957078.1 | 1376 | 61 | ||
| XM_021101420.1 | XP_020957079.1 | 789 | 61 | |||
| XM_021101421.1 | XP_020957080.1 | 1100 | 61 | |||
| XM_021101422.1 | XP_020957081.1 | 880 | 61 | |||
| GenBank mRNA | HQ681285.1 | - | 1 | 116 | 61 | |
More Information
Set Membership: |
|

NPFFR2
Genomic Map