Assay Details
Gene Symbol: HS2ST1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001925492.6 | XP_001925527.3 | 5839 | 93 | ||
XM_013997435.2 | XP_013852889.2 | 5933 | 93 | |||
XM_013997436.2 | XP_013852890.2 | 5727 | 93 | |||
GenBank mRNA | AK233667.1 | - | 1 | 2620 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
