Assay Details
| Gene Symbol: MEPCE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003124352.6 | XP_003124400.1 | 2735 | 66 | ||
| GenBank mRNA | AK233695.1 | - | 1 - 2 | 990 | 66 | |
| AK236581.1 | - | 1 - 2 | 927 | 66 | ||
| AK351554.1 | - | 1 - 2 | 1954 | 66 | ||
More Information
Set Membership: |
|

MEPCE
Genomic Map