Assay Details
| Gene Symbol: IL21R | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021086442.1 | XP_020942101.1 | 1338 | 77 | ||
| XM_021086443.1 | XP_020942102.1 | 793 | 77 | |||
| GenBank mRNA | AK233853.1 | - | 7 - 8 | 938 | 77 | |
More Information
Set Membership: |
|

IL21R
Genomic Map