Assay Details
| Gene Symbol: SEC11C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003121713.3 | XP_003121761.3 | 534 | 84 | ||
| GenBank mRNA | AK233891.1 | - | 1 - 2 | 142 | 84 | |
| AK391750.1 | - | 1 - 2 | 198 | 84 | ||
| AK392595.1 | - | 1 - 2 | 152 | 84 | ||
| AK400074.1 | - | 1 - 2 | 439 | 84 | ||
| AY610341.1 | - | 1 - 2 | 128 | 84 | ||
More Information
Set Membership: |
|

SEC11C
Genomic Map