Assay Details
| Gene Symbol: TMEM170A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003126863.6 | XP_003126911.3 | 290 | 61 | ||
| XM_005664341.3 | XP_005664398.1 | 292 | 61 | |||
| XM_013998173.2 | XP_013853627.1 | 294 | 61 | |||
| GenBank mRNA | AK233991.1 | - | 1 - 2 | 280 | 61 | |
More Information
Set Membership: |
|

TMEM170A
Genomic Map