Assay Details
| Gene Symbol: METTL26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021086912.1 | XP_020942571.1 | 289 | 80 | ||
| GenBank mRNA | AK233999.1 | - | 1 - 2 | 198 | 80 | |
More Information
Set Membership: |
|

METTL26
Genomic Map