Assay Details
Gene Symbol: C1H18orf32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK234089.1 | - | 2 - 3 | 422 | 89 | |
AK235845.1 | - | 2 - 3 | 437 | 89 | ||
AK392356.1 | - | 2 - 3 | 474 | 89 | ||
AK399944.1 | - | 2 - 3 | 309 | 89 | ||
AK400750.1 | - | 2 - 3 | 424 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
