Assay Details
| Gene Symbol: DET1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005660034.3 | XP_005660091.1 | 1617 | 82 | ||
| XM_005660035.3 | XP_005660092.1 | 1607 | 82 | |||
| XM_013993479.2 | XP_013848933.1 | 1674 | 82 | |||
| GenBank mRNA | AK234221.1 | - | 4 - 5 | 1581 | 82 | |
| AK234419.1 | - | 5 - 6 | 1528 | 82 | ||
More Information
Set Membership: |
|

DET1
Genomic Map