Assay Details
Gene Symbol: LOC100157391 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001926555.6 | XP_001926590.3 | 347 | 104 | ||
XM_005671220.3 | XP_005671277.1 | 272 | 104 | |||
GenBank mRNA | AK234276.1 | - | 1 - 2 | 229 | 104 | |
AK235825.1 | - | 1 - 2 | 270 | 104 | ||
AK392422.1 | - | 1 - 2 | 234 | 104 | ||
AK400657.1 | - | 1 - 2 | 275 | 104 | ||
AY610146.1 | - | 1 - 2 | 352 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
