Assay Details
Gene Symbol: LOC100157391 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001926555.6 | XP_001926590.3 | 403 | 99 | ||
XM_005671220.3 | XP_005671277.1 | 328 | 99 | |||
GenBank mRNA | AK234276.1 | - | 2 - 3 | 285 | 99 | |
AK235825.1 | - | 2 - 3 | 326 | 99 | ||
AK392422.1 | - | 2 - 3 | 290 | 99 | ||
AK400657.1 | - | 2 - 3 | 331 | 99 | ||
AY610146.1 | - | 2 - 3 | 408 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
