Assay Details
Gene Symbol: LOC100153598 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021068951.1 | XP_020924610.1 | 290 | 63 | ||
GenBank mRNA | AK234441.1 | - | 1 - 2 | 297 | 63 | |
AK350584.1 | - | 1 - 2 | 265 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() |
