Assay Details
Gene Symbol: LOC100153598 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021068950.1 | XP_020924609.1 | 672 | 128 | ||
XM_021068951.1 | XP_020924610.1 | 560 | 128 | |||
GenBank mRNA | AK234441.1 | - | 4 - 5 | 567 | 128 | |
AK350584.1 | - | 4 - 5 | 535 | 128 |

More Information
Set Membership: |
![]() ![]() ![]() |
