Assay Details
Gene Symbol: LOC100153598 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021068950.1 | XP_020924609.1 | 787 | 71 | ||
XM_021068951.1 | XP_020924610.1 | 675 | 71 | |||
GenBank mRNA | AK234441.1 | - | 5 - 6 | 682 | 71 | |
AK350584.1 | - | 5 - 6 | 650 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() |
