Assay Details
Gene Symbol: LOC100153598 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021068950.1 | XP_020924609.1 | 1013 | 106 | ||
XM_021068951.1 | XP_020924610.1 | 901 | 106 | |||
GenBank mRNA | AK234441.1 | - | 7 - 8 | 908 | 106 | |
AK350584.1 | - | 7 - 8 | 876 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
