Assay Details
| Gene Symbol: EID1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001928761.4 | XP_001928796.1 | 1422 | 100 | ||
| GenBank mRNA | AK235101.1 | - | 1 | 1119 | 100 | |
| AK392040.1 | - | 1 | 1142 | 100 | ||
More Information
Set Membership: |
|

EID1
Genomic Map