Assay Details
Gene Symbol: CDYL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021100128.1 | XP_020955787.1 | 1865 | 73 | ||
XM_021100129.1 | XP_020955788.1 | 1648 | 73 | |||
XM_021100130.1 | XP_020955789.1 | 1865 | 73 | |||
GenBank mRNA | AK235146.1 | - | 2 - 3 | 355 | 73 | |
AK345435.1 | - | 2 - 3 | 285 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
