Assay Details
| Gene Symbol: MLEC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001928694.6 | XP_001928729.1 | 6071 | 86 | ||
| GenBank mRNA | AK235220.1 | - | 1 | 1222 | 86 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: MLEC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001928694.6 | XP_001928729.1 | 6071 | 86 | ||
| GenBank mRNA | AK235220.1 | - | 1 | 1222 | 86 | |
Set Membership: |
|