Assay Details
| Gene Symbol: TRAPPC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005665219.3 | XP_005665276.1 | 392 | 66 | ||
| GenBank mRNA | AK235322.1 | - | 3 - 4 | 356 | 66 | |
| AK352016.1 | - | 3 - 4 | 351 | 66 | ||
| AK389506.1 | - | 3 - 4 | 370 | 66 | ||
More Information
Set Membership: |
|

TRAPPC3
Genomic Map