Assay Details
| Gene Symbol: RPS8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003128060.4 | XP_003128108.1 | 299 | 94 | ||
| GenBank mRNA | AK235428.1 | - | 3 - 4 | 240 | 94 | |
| AK392546.1 | - | 3 - 4 | 241 | 94 | ||
More Information
Set Membership: |
|

RPS8
Genomic Map