Assay Details
Gene Symbol: IDS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021080476.1 | XP_020936135.1 | 873 | 61 | ||
XM_021080477.1 | XP_020936136.1 | 873 | 61 | |||
XM_021080478.1 | XP_020936137.1 | 873 | 61 | |||
GenBank mRNA | AK235784.1 | - | 5 - 6 | 860 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
