Assay Details
| Gene Symbol: RRM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021087849.1 | XP_020943508.1 | 1917 | 69 | ||
| XM_021087850.1 | XP_020943509.1 | 1043 | 69 | |||
| GenBank mRNA | AB530148.1 | - | 7 - 8 | 750 | 69 | |
| AK239473.1 | - | 7 - 8 | 848 | 69 | ||
More Information
Set Membership: |
|

RRM2
Genomic Map