Assay Details
Gene Symbol: CIAO2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021093917.1 | XP_020949576.1 | 507 | 94 | ||
GenBank mRNA | AK230508.1 | - | 3 - 4 | 352 | 94 | |
AK394232.1 | - | 3 - 4 | 417 | 94 | ||
AK399759.1 | - | 3 - 4 | 470 | 94 | ||
AY610212.1 | - | 3 - 4 | 468 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() |
