Assay Details
| Gene Symbol: BLZF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003355067.4 | XP_003355115.1 | 891 | 71 | ||
| GenBank mRNA | AK230581.1 | - | 3 - 4 | 660 | 71 | |
| AK230832.1 | - | 3 - 4 | 660 | 71 | ||
| AK391474.1 | - | 3 - 4 | 71 | |||
More Information
Set Membership: |
|

BLZF1
Genomic Map