Assay Details
Gene Symbol: IPCEF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021086620.1 | XP_020942279.1 | 2321 | 84 | ||
GenBank mRNA | AK230687.1 | - | 8 - 9 | 699 | 84 | |
AK389763.1 | - | 4 - 5 | 519 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
