Assay Details
| Gene Symbol: LOC100156062 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005663839.3 | XP_005663896.2 | 4255 | 83 | ||
| XM_013997551.2 | XP_013853005.2 | 4297 | 83 | |||
| XM_021091539.1 | XP_020947198.1 | 4962 | 83 | |||
| XM_021091540.1 | XP_020947199.1 | 4152 | 83 | |||
| XM_021091541.1 | XP_020947200.1 | 4178 | 83 | |||
| XM_021091542.1 | XP_020947201.1 | 4032 | 83 | |||
| GenBank mRNA | AK230871.1 | - | 1 | 1921 | 84 | |
More Information
Set Membership: |
|

LOC100156062
Genomic Map