Assay Details
Gene Symbol: C14H12orf43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001929089.4 | XP_001929124.1 | 267 | 94 | ||
GenBank mRNA | AK235787.1 | - | 2 - 3 | 218 | 94 | |
AK238156.1 | - | 2 - 3 | 219 | 94 | ||
AK346318.1 | - | 2 - 3 | 213 | 94 | ||
AK398027.1 | - | 2 - 3 | 220 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() |
