Assay Details
Gene Symbol: C14H12orf43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001929089.4 | XP_001929124.1 | 357 | 71 | ||
GenBank mRNA | AK235787.1 | - | 3 - 4 | 308 | 71 | |
AK238156.1 | - | 3 - 4 | 309 | 71 | ||
AK346318.1 | - | 3 - 4 | 303 | 71 | ||
AK398027.1 | - | 3 - 4 | 310 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() |
