Assay Details
Gene Symbol: C14H12orf43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001929089.4 | XP_001929124.1 | 433 | 87 | ||
GenBank mRNA | AK235787.1 | - | 4 - 5 | 384 | 87 | |
AK238156.1 | - | 4 - 5 | 385 | 87 | ||
AK346318.1 | - | 4 - 5 | 379 | 87 | ||
AK398027.1 | - | 4 - 5 | 386 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() |
