Assay Details
Gene Symbol: TSLP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021085155.1 | XP_020940814.1 | 442 | 89 | ||
GenBank mRNA | AK235867.1 | - | 2 - 3 | 657 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
