Assay Details
Gene Symbol: TSLP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021085155.1 | XP_020940814.1 | 614 | 83 | ||
GenBank mRNA | AK235867.1 | - | 4 - 5 | 829 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
