Assay Details
| Gene Symbol: HSPA13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001924920.6 | XP_001924955.1 | 678 | 97 | ||
| GenBank mRNA | AK235890.1 | - | 3 - 4 | 579 | 97 | |
| AK389713.1 | - | 3 - 4 | 592 | 97 | ||
More Information
Set Membership: |
|

HSPA13
Genomic Map