Assay Details
| Gene Symbol: KTI12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021096660.1 | XP_020952319.1 | 1673 | 68 | ||
| GenBank mRNA | AK236002.1 | - | 1 | 626 | 68 | |
| AK239449.1 | - | 1 | 628 | 68 | ||
More Information
Set Membership: |
|

KTI12
Genomic Map