Assay Details
Gene Symbol: DFFA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003127563.4 | XP_003127611.1 | 710 | 69 | ||
GenBank mRNA | AK236061.1 | - | 4 - 5 | 646 | 69 | |
AK390962.1 | - | 4 - 5 | 715 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
