Assay Details
Gene Symbol: MESD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001928863.5 | XP_001928898.1 | 569 | 64 | ||
GenBank mRNA | AK236081.1 | - | 2 - 3 | 471 | 64 | |
AK392695.1 | - | 2 - 3 | 456 | 64 | ||
AK396798.1 | - | 2 - 3 | 458 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
