Assay Details
| Gene Symbol: NSRP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003131784.6 | XP_003131832.1 | 697 | 110 | ||
| XM_013981200.2 | XP_013836654.1 | 556 | 110 | |||
| XM_021067519.1 | XP_020923178.1 | 782 | 110 | |||
| GenBank mRNA | AK236102.1 | - | 6 - 7 | 664 | 110 | |
| AK391549.1 | - | 7 - 8 | 783 | 110 | ||
| AK392117.1 | - | 6 - 7 | 647 | 110 | ||
More Information
Set Membership: |
|

NSRP1
Genomic Map