Assay Details
Gene Symbol: NONO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK236143.1 | - | 9 - 10 | 1249 | 56 | |
AK238771.1 | - | 3 - 4 | 240 | 56 | ||
AK352015.1 | - | 9 - 10 | 1283 | 56 | ||
AK394102.1 | - | 9 - 10 | 1416 | 56 | ||
AK395168.1 | - | 9 - 10 | 1243 | 56 | ||
AK399131.1 | - | 9 - 10 | 1247 | 56 | ||
AK399282.1 | - | 8 - 9 | 1097 | 56 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
