Assay Details
Gene Symbol: TESK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003121984.4 | XP_003122032.1 | 1099 | 80 | ||
XM_013993645.2 | XP_013849099.1 | 708 | 80 | |||
XM_021065633.1 | XP_020921292.1 | 313 | 80 | |||
GenBank mRNA | AK236260.1 | - | 3 - 4 | 226 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
