Assay Details
Gene Symbol: YEATS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021091973.1 | XP_020947632.1 | 757 | 80 | ||
XM_021091975.1 | XP_020947634.1 | 743 | 80 | |||
GenBank mRNA | AK236353.1 | - | 6 - 7 | 745 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
